Assay Details
| Gene Symbol: GALNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291866.1 | NP_001278795.1 | 10 - 11 | 1180 | 84 | |
| NM_004481.4 | NP_004472.1 | 10 - 11 | 1102 | 84 | ||
| XM_017000963.1 | XP_016856452.1 | 10 - 11 | 1062 | 84 | ||
| XM_017000964.1 | XP_016856453.1 | 10 - 11 | 1687 | 84 | ||
| XM_017000965.1 | XP_016856454.1 | 10 - 11 | 1180 | 84 | ||
| XM_017000966.1 | XP_016856455.1 | 10 - 11 | 1005 | 84 | ||
| GenBank mRNA | AK290048.1 | - | 10 - 11 | 1028 | 84 | |
| AK296886.1 | - | 7 - 8 | 699 | 84 | ||
| AK300453.1 | - | 10 - 11 | 1180 | 84 | ||
| BC041120.1 | - | 10 - 11 | 1088 | 5553465 | 84 | |
| X85019.1 | - | 10 - 11 | 1016 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001907.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
immunoglobulin biosynthetic process
protein O-linked glycosylation
O-glycan processing
protein O-linked glycosylation via serine
protein O-linked glycosylation via threonine
polypeptide N-acetylgalactosaminyltransferase activity
protein binding
manganese ion binding
carbohydrate binding
immunoglobulin biosynthetic process
protein O-linked glycosylation
O-glycan processing
protein O-linked glycosylation via serine
protein O-linked glycosylation via threonine
polypeptide N-acetylgalactosaminyltransferase activity
protein binding
manganese ion binding
carbohydrate binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000488903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000488903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029617.hg.1 | TC01001907.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029617.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029617.hg.1 | ENST00000541865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029617.hg.1 | ENST00000543291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029617.hg.1 | ENST00000543760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029617.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000488903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000488903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187970.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100187970.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187970.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187970.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100187970.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187970.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187970.hg.1 | TC0100011920.hg.1 | Clariom D TC |

GALNT2
Genomic Map