Assay Details
| Gene Symbol: GALNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291866.1 | NP_001278795.1 | 1 - 2 | 291 | 100 | |
| XM_017000965.1 | XP_016856454.1 | 1 - 2 | 291 | 100 | ||
| GenBank mRNA | AK300453.1 | - | 1 - 2 | 291 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK300453.1, NM_001291866.1, XM_017000965.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
immunoglobulin biosynthetic process
protein O-linked glycosylation
O-glycan processing
protein O-linked glycosylation via serine
protein O-linked glycosylation via threonine
polypeptide N-acetylgalactosaminyltransferase activity
protein binding
manganese ion binding
carbohydrate binding
immunoglobulin biosynthetic process
protein O-linked glycosylation
O-glycan processing
protein O-linked glycosylation via serine
protein O-linked glycosylation via threonine
polypeptide N-acetylgalactosaminyltransferase activity
protein binding
manganese ion binding
carbohydrate binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000488903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000488903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187948.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187948.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187948.hg.1 | TC0100011920.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100187957.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100187957.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100187957.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187957.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187957.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100187957.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100187957.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187957.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187957.hg.1 | TC0100011920.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000488903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000488903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187949.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100187949.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187949.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100187949.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187949.hg.1 | TC0100011920.hg.1 | Clariom D TC |

GALNT2
Genomic Map