Assay Details
| Gene Symbol: PALB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024675.3 | NP_078951.2 | 10 - 11 | 3308 | 129 | |
| XM_011545946.2 | XP_011544248.1 | 10 - 11 | 4113 | 129 | ||
| XM_011545947.2 | XP_011544249.1 | 10 - 11 | 4078 | 129 | ||
| XM_011545948.2 | XP_011544250.1 | 11 - 12 | 3400 | 129 | ||
| XM_017023673.1 | XP_016879162.1 | 10 - 11 | 3308 | 129 | ||
| GenBank mRNA | AK025469.1 | - | 7 - 8 | 1685 | 129 | |
| AK299152.1 | - | 10 - 11 | 3205 | 129 | ||
| AL834425.1 | - | 7 - 8 | 2591 | 129 | ||
| BC044254.2 | - | 10 - 11 | 3235 | 6045564 | 129 | |
| BX647125.1 | - | 10 - 11 | 3219 | 129 | ||
| BX647130.1 | - | 10 - 11 | 3261 | 129 | ||
| CR749637.1 | - | 10 - 11 | 3259 | 129 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000967.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
DNA synthesis involved in DNA repair
strand displacement
somitogenesis
inner cell mass cell proliferation
mesoderm development
organ morphogenesis
chromosome breakage
multicellular organism growth
post-anal tail morphogenesis
negative regulation of apoptotic process
embryonic organ development
DNA binding
protein binding
double-strand break repair via homologous recombination
DNA synthesis involved in DNA repair
strand displacement
somitogenesis
inner cell mass cell proliferation
mesoderm development
organ morphogenesis
chromosome breakage
multicellular organism growth
post-anal tail morphogenesis
negative regulation of apoptotic process
embryonic organ development
DNA binding
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000561514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000561514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000561514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000561514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000565038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000565038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000565038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000566069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000566069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000566069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000566069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000567003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000567003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000567003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000567003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000567003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000567003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000567003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013912.hg.1 | ENST00000261584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013912.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013912.hg.1 | TC16000967.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600175105.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175105.hg.1 | ENST00000566069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175105.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175105.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175105.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175105.hg.1 | ENST00000566069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175105.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175105.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175105.hg.1 | TC1600009746.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600175107.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175107.hg.1 | ENST00000566069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175107.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175107.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175107.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175107.hg.1 | ENST00000566069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175107.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175107.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175107.hg.1 | TC1600009746.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000261584.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000561514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000561514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000561514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000561514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000565038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000565038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000565038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000566069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000566069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000566069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000566069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000567003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000567003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000567003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000567003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000567003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000567003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000567003.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | ENST00000568219.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009746.hg.1 | NM_024675.3 | Refseq |

PALB2
Genomic Map