Assay Details
| Gene Symbol: MRVI1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046374.1 | 2 - 3 | 401 | 62 | ||
| GenBank mRNA | BI520309.1 | - | 2 - 3 | 340 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000525578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000525578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000525578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000525578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000525578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000525578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_034093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_034093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_034093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_034093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_034093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_034093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_034093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_034094.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_034094.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_034094.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_034094.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_034094.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_034094.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148086.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148086.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148086.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148086.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148086.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148086.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148086.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148086.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148086.hg.1 | TC1100006836.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000525578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000525578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000525578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000525578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000525578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000525578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | ENST00000529979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_034093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_034093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_034093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_034093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_034093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_034093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_034093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_034094.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_034094.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_034094.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_034094.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_034094.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_034094.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046374.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006836.hg.1 | NR_046375.1 | Refseq |

MRVI1-AS1
Genomic Map