Assay Details
| Gene Symbol: ADRA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033302.3 | NP_150645.2 | 2 - 3 | 1956 | 96 | |
| XM_006716293.3 | XP_006716356.1 | 3 - 4 | 2291 | 96 | ||
| GenBank mRNA | D32201.1 | - | 2 - 3 | 1706 | 96 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001076.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of heart rate involved in baroreceptor response to increased systemic arterial blood pressure
norepinephrine-epinephrine vasoconstriction involved in regulation of systemic arterial blood pressure
positive regulation of heart rate by epinephrine-norepinephrine
positive regulation of the force of heart contraction by epinephrine-norepinephrine
positive regulation of systemic arterial blood pressure
apoptotic process
smooth muscle contraction
signal transduction
G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
activation of phospholipase C activity
positive regulation of cytosolic calcium ion concentration
cell-cell signaling
adult heart development
aging
negative regulation of cell proliferation
response to hormone
cell growth
positive regulation of synaptic transmission, GABAergic
negative regulation of Rho protein signal transduction
organ growth
intracellular signal transduction
response to drug
positive regulation of MAPK cascade
positive regulation of action potential
positive regulation of vasoconstriction
positive regulation of smooth muscle contraction
micturition
calcium ion transport into cytosol
positive regulation of cardiac muscle contraction
positive regulation of ERK1 and ERK2 cascade
adenylate cyclase-activating adrenergic receptor signaling pathway
positive regulation of protein kinase C signaling
pilomotor reflex
alpha1-adrenergic receptor activity
protein heterodimerization activity
negative regulation of heart rate involved in baroreceptor response to increased systemic arterial blood pressure
norepinephrine-epinephrine vasoconstriction involved in regulation of systemic arterial blood pressure
positive regulation of heart rate by epinephrine-norepinephrine
positive regulation of the force of heart contraction by epinephrine-norepinephrine
positive regulation of systemic arterial blood pressure
apoptotic process
smooth muscle contraction
signal transduction
G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
activation of phospholipase C activity
positive regulation of cytosolic calcium ion concentration
cell-cell signaling
adult heart development
aging
negative regulation of cell proliferation
response to hormone
cell growth
positive regulation of synaptic transmission, GABAergic
negative regulation of Rho protein signal transduction
organ growth
intracellular signal transduction
response to drug
positive regulation of MAPK cascade
positive regulation of action potential
positive regulation of vasoconstriction
positive regulation of smooth muscle contraction
micturition
calcium ion transport into cytosol
positive regulation of cardiac muscle contraction
positive regulation of ERK1 and ERK2 cascade
adenylate cyclase-activating adrenergic receptor signaling pathway
positive regulation of protein kinase C signaling
pilomotor reflex
alpha1-adrenergic receptor activity
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000276393.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000276393.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000276393.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000276393.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000276393.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000276393.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000276393.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000354550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000354550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000354550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000354550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000354550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000380572.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000380572.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000380572.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000380572.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000380582.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000380582.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000380582.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000380582.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000380582.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000380582.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000380586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000380586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000380586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000380586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000380586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000380586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000518621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000518621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000518621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000518621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000518810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000518810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000518810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000518810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000518810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000519096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000519096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000519096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000519096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000519096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000519096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000519229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000519229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000519229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000519229.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000521711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000521711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000521711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000521711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000521711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | ENST00000521711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_000680.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_000680.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_000680.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_000680.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_000680.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_000680.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_000680.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033302.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033303.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033303.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033303.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033303.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033303.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033303.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033304.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033304.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033304.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033304.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033304.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012393.hg.1 | NM_033304.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013699.hg.1 | TC08001076.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013699.hg.1 | ENST00000380582.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013699.hg.1 | NM_033302.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186799.hg.1 | ENST00000380582.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186799.hg.1 | ENST00000521711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186799.hg.1 | NM_033302.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186799.hg.1 | ENST00000380582.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186799.hg.1 | ENST00000521711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186799.hg.1 | NM_033302.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186799.hg.1 | TC0800012393.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000276393.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000276393.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000276393.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000276393.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000276393.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000276393.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000276393.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000354550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000354550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000354550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000354550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000354550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000380572.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000380572.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000380572.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000380572.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000380582.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000380582.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000380582.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000380582.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000380582.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000380582.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000380586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000380586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000380586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000380586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000380586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000380586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000518621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000518621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000518621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000518621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000518810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000518810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000518810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000518810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000518810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000519096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000519096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000519096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000519096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000519096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000519096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000519229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000519229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000519229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000519229.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000521711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000521711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000521711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000521711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000521711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | ENST00000521711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_000680.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_000680.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_000680.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_000680.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_000680.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_000680.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_000680.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033302.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033303.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033303.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033303.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033303.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033303.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033303.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033304.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033304.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033304.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033304.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033304.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012393.hg.1 | NM_033304.3 | Refseq |

ADRA1A
Genomic Map