Assay Details
| Gene Symbol: CD1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042585.2 | NP_001036050.1 | 4 - 5 | 1032 | 70 | |
| NM_001185108.1 | NP_001172037.1 | 3 - 4 | 762 | 70 | ||
| NM_001185110.1 | NP_001172039.1 | 2 - 3 | 465 | 70 | ||
| NM_001185113.1 | NP_001172042.1 | 2 - 3 | 465 | 70 | ||
| XM_011510133.2 | XP_011508435.1 | 4 - 5 | 1571 | 70 | ||
| GenBank mRNA | AJ289114.1 | - | 4 - 5 | 739 | 70 | |
| AJ289117.1 | - | 3 - 4 | 469 | 70 | ||
| AJ289121.1 | - | 2 - 3 | 172 | 70 | ||
| AJ289122.1 | - | 2 - 3 | 172 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001340.hg.1
Set Membership: |
|

CD1E
Genomic Map