Assay Details
| Gene Symbol: GPR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF258342.1 | - | 2 - 3 | 257 | 149 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF258342.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000526744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000526744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000526744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000404129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000469383.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000526744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000526744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000526744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000527748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000533959.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000616874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | ENST00000618721.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009355.hg.1 | NM_031936.4 | Refseq |

GPR61
Genomic Map