Assay Details
| Gene Symbol: GPBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006714673.3 | XP_006714736.1 | 8 - 9 | 1888 | 87 | |
| XM_006714674.3 | XP_006714737.1 | 7 - 8 | 1867 | 87 | ||
| XM_017009754.1 | XP_016865243.1 | 7 - 8 | 1241 | 87 | ||
| XM_017009755.1 | XP_016865244.1 | 6 - 7 | 1220 | 87 | ||
| GenBank mRNA | AK307505.1 | - | 4 - 5 | 405 | 87 | |
| CD657101.1 | - | 4 - 5 | 401 | 87 | ||
More Information
Set Membership: |
|

GPBP1
Genomic Map