Assay Details
| Gene Symbol: KRT25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181534.3 | NP_853512.1 | 6 - 7 | 1234 | 123 | |
| XM_011524414.1 | XP_011522716.1 | 7 - 8 | 1222 | 123 | ||
| GenBank mRNA | AJ564204.1 | - | 6 - 7 | 1215 | 123 | |
| AK129503.1 | - | 6 - 7 | 1036 | 123 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17002915.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | ENST00000540723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | ENST00000540723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | ENST00000540723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | ENST00000540723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | ENST00000540723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019416.hg.1 | TC17002915.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019416.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019416.hg.1 | ENST00000394042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019416.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019415.hg.1 | TC17002915.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019415.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019415.hg.1 | ENST00000394042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019415.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019417.hg.1 | TC17002915.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019417.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019417.hg.1 | ENST00000394042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019417.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206253.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206253.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206253.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206253.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206253.hg.1 | TC1700012397.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012397.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206254.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206254.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206254.hg.1 | ENST00000312150.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700206254.hg.1 | NM_181534.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700206254.hg.1 | TC1700012397.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | ENST00000301656.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | ENST00000540723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | ENST00000540723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | ENST00000540723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | ENST00000540723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | ENST00000540723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | NM_181537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012398.hg.1 | NM_181537.3 | Refseq |

KRT25
Genomic Map