Assay Details
Gene Symbol: DPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003859.2 | NP_003850.1 | 6 - 7 | 537 | 84 | |
GenBank mRNA | AK289569.1 | - | 6 - 7 | 505 | 84 | |
BC007073.1 | - | 6 - 7 | 523 | 3997453 | 84 | |
BC008466.1 | - | 6 - 7 | 523 | 4276338 | 84 | |
BC016322.1 | - | 6 - 7 | 508 | 4083041 | 84 | |
CR456926.1 | - | 6 - 7 | 496 | 84 | ||
D86198.1 | - | 6 - 7 | 496 | 84 | ||
KU178444.1 | - | 6 - 7 | 496 | 84 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000940.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein N-linked glycosylation
GPI anchor biosynthetic process
protein N-linked glycosylation via asparagine
dolichol metabolic process
GDP-mannose metabolic process
protein mannosylation
protein O-linked mannosylation
dolichyl-phosphate-mannose-protein mannosyltransferase activity
dolichyl-phosphate beta-D-mannosyltransferase activity
protein binding
mannose binding
alcohol binding
protein N-linked glycosylation
GPI anchor biosynthetic process
protein N-linked glycosylation via asparagine
dolichol metabolic process
GDP-mannose metabolic process
protein mannosylation
protein O-linked mannosylation
dolichyl-phosphate-mannose-protein mannosyltransferase activity
dolichyl-phosphate beta-D-mannosyltransferase activity
protein binding
mannose binding
alcohol binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR20012954.hg.1 | ENST00000371582.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20012954.hg.1 | ENST00000371583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20012954.hg.1 | ENST00000371588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20012954.hg.1 | ENST00000449701.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20012954.hg.1 | ENST00000466152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20012954.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20012954.hg.1 | TC20000940.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20012955.hg.1 | ENST00000371582.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20012955.hg.1 | ENST00000371583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20012955.hg.1 | ENST00000371588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20012955.hg.1 | ENST00000449701.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20012955.hg.1 | ENST00000466152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20012955.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20012955.hg.1 | TC20000940.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009447.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000166060.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000166060.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000166060.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000166060.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000166060.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000166060.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000166060.hg.1 | NM_003859.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000166060.hg.1 | TC2000009447.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000166060.hg.1 | ENST00000371582.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000166060.hg.1 | ENST00000371584.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000166060.hg.1 | ENST00000371588.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000166060.hg.1 | ENST00000413082.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000166060.hg.1 | ENST00000466152.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000166060.hg.1 | ENST00000494752.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000166060.hg.1 | NM_003859.2 | Refseq |
