Assay Details
| Gene Symbol: PDS5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100399.1 | NP_001093869.1 | 22 - 23 | 3048 | 67 | |
| XM_011513672.2 | XP_011511974.1 | 22 - 23 | 2661 | 67 | ||
| XM_017007928.1 | XP_016863417.1 | 22 - 23 | 2623 | 67 | ||
| GenBank mRNA | AB014548.1 | - | 10 - 11 | 1052 | 67 | |
| AB621803.1 | - | 22 - 23 | 3062 | 67 | ||
| AF294791.1 | - | 22 - 23 | 2619 | 67 | ||
| BC114218.1 | - | 22 - 23 | 2744 | 5579832 | 67 | |
| BC126225.1 | - | 21 - 22 | 2458 | 8991941 | 67 | |
| CR749425.1 | - | 21 - 22 | 2417 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001134.hg.1
Set Membership: |
|

PDS5A
Genomic Map