Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
ENST00000267512.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
ENST00000426039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
ENST00000533601.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
ENST00000554593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
ENST00000555256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
ENST00000585059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
NM_001308154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
NM_198686.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
ENST00000267512.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
ENST00000426039.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
ENST00000533601.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
ENST00000554593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
ENST00000555256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
ENST00000585059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
NM_001308154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
NM_198686.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169639.hg.1 |
TC1400009422.hg.1 |
Clariom D TC |