Assay Details
| Gene Symbol: MDN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014611.2 | NP_055426.1 | 41 - 42 | 6247 | 112 | |
| XM_005248700.3 | XP_005248757.1 | 2 - 3 | 179 | 112 | ||
| XM_006715405.3 | XP_006715468.1 | 5 - 6 | 1725 | 112 | ||
| XM_011535635.2 | XP_011533937.1 | 41 - 42 | 6052 | 112 | ||
| XM_011535636.2 | XP_011533938.1 | 41 - 42 | 6217 | 112 | ||
| GenBank mRNA | AF503925.1 | - | 41 - 42 | 6176 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005248700.3 transcripts.
Set Membership: |
|

MDN1
Genomic Map