Assay Details
| Gene Symbol: MDN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014611.2 | NP_055426.1 | 47 - 48 | 7402 | 97 | |
| XM_005248700.3 | XP_005248757.1 | 8 - 9 | 1334 | 97 | ||
| XM_006715405.3 | XP_006715468.1 | 11 - 12 | 2880 | 97 | ||
| XM_011535635.2 | XP_011533937.1 | 47 - 48 | 7207 | 97 | ||
| XM_011535636.2 | XP_011533938.1 | 47 - 48 | 7372 | 97 | ||
| GenBank mRNA | AB002299.2 | - | 1 - 2 | 58 | 97 | |
| AF503925.1 | - | 47 - 48 | 7331 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

MDN1
Genomic Map