| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013188.hg.1 |
ENST00000371435.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013188.hg.1 |
ENST00000371440.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013188.hg.1 |
ENST00000395961.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013188.hg.1 |
ENST00000434986.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013188.hg.1 |
ENST00000448710.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013188.hg.1 |
NM_003657.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013188.hg.1 |
TC20000960.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013189.hg.1 |
ENST00000371435.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013189.hg.1 |
ENST00000371440.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013189.hg.1 |
ENST00000395961.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013189.hg.1 |
ENST00000434986.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013189.hg.1 |
ENST00000448710.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013189.hg.1 |
NM_003657.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013189.hg.1 |
TC20000960.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013186.hg.1 |
ENST00000371435.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013186.hg.1 |
ENST00000371440.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013186.hg.1 |
ENST00000395961.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013186.hg.1 |
ENST00000434986.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013186.hg.1 |
ENST00000448710.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013186.hg.1 |
NM_003657.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013186.hg.1 |
TC20000960.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2000166496.hg.1 |
ENST00000371435.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166496.hg.1 |
ENST00000371440.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166496.hg.1 |
ENST00000395961.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166496.hg.1 |
ENST00000422805.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166496.hg.1 |
ENST00000448484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166496.hg.1 |
NM_003657.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000166496.hg.1 |
TC2000009516.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000166496.hg.1 |
ENST00000371435.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166496.hg.1 |
ENST00000371440.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166496.hg.1 |
ENST00000395961.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166496.hg.1 |
ENST00000422805.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166496.hg.1 |
ENST00000448484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166496.hg.1 |
NM_003657.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000166498.hg.1 |
ENST00000371435.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166498.hg.1 |
ENST00000371440.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166498.hg.1 |
ENST00000395961.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166498.hg.1 |
ENST00000422805.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166498.hg.1 |
ENST00000448484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166498.hg.1 |
NM_003657.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000166498.hg.1 |
TC2000009516.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000166498.hg.1 |
ENST00000371435.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166498.hg.1 |
ENST00000371440.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166498.hg.1 |
ENST00000395961.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166498.hg.1 |
ENST00000422805.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166498.hg.1 |
ENST00000448484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166498.hg.1 |
NM_003657.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000166499.hg.1 |
ENST00000371435.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166499.hg.1 |
ENST00000371440.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166499.hg.1 |
ENST00000395961.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166499.hg.1 |
ENST00000422805.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166499.hg.1 |
ENST00000448484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166499.hg.1 |
NM_003657.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000166499.hg.1 |
TC2000009516.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000166499.hg.1 |
ENST00000371435.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166499.hg.1 |
ENST00000371440.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166499.hg.1 |
ENST00000395961.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166499.hg.1 |
ENST00000422805.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166499.hg.1 |
ENST00000448484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166499.hg.1 |
NM_003657.3 |
Refseq |