Assay Details
| Gene Symbol: CBFA2T2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005093.3 | NP_005084.1 | 2 - 3 | 599 | 52 | |
| GenBank mRNA | AF013970.1 | - | 1 - 2 | 72 | 52 | |
| AK314159.1 | - | 2 - 3 | 398 | 52 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000231.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
positive regulation of neuron projection development
negative regulation of neuron projection development
epithelial cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
metal ion binding
transcription, DNA-templated
positive regulation of neuron projection development
negative regulation of neuron projection development
epithelial cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000342704.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000342704.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000342704.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000342704.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000342704.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000342704.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000342704.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000342704.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000342704.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000342704.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000342704.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000342704.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000342704.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000342704.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000342704.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000344201.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000344201.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000344201.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000344201.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000344201.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000344201.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000344201.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000344201.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000344201.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000346541.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000359606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000375279.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000397800.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000471007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000471007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000471007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000471007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000471007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000471007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000491618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000491618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000491618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000491618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000491618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000491618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000492345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000543126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000543126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000543126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000543126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000543126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | ENST00000543126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001032999.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_001039709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007157.hg.1 | NM_005093.3 | Refseq |

CBFA2T2
Genomic Map