Assay Details
Gene Symbol: FKBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004470.3 | NP_004461.2 | 1 - 2 | 198 | 138 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000590.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_004470.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000449942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000449942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000449942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000449942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000449942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000449942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000449942.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000535135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000535135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000535135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000535135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000536642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000536642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000536642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000536642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000536642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000536642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_001135208.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_001135208.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_001135208.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_001135208.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_001135208.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_001135208.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_001135208.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_057092.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_057092.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_057092.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_057092.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_057092.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_057092.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007941.hg.1 | NM_057092.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11007200.hg.1 | ENST00000309366.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11007200.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11007200.hg.1 | TC11000590.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11007198.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11007198.hg.1 | TC11000590.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11007199.hg.1 | ENST00000309366.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11007199.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11007199.hg.1 | TC11000590.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100156831.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100156831.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100156831.hg.1 | TC1100007941.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100156833.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100156833.hg.1 | ENST00000536642.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100156833.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100156833.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100156833.hg.1 | ENST00000536642.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100156833.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100156833.hg.1 | TC1100007941.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000394540.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000449942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000449942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000449942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000449942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000449942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000449942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000449942.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000535135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000535135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000535135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000535135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000536642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000536642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000536642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000536642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000536642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000536642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_001135208.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_001135208.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_001135208.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_001135208.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_001135208.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_001135208.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_001135208.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_057092.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_057092.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_057092.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_057092.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_057092.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_057092.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007941.hg.1 | NM_057092.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100156835.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100156835.hg.1 | ENST00000536642.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100156835.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100156835.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100156835.hg.1 | ENST00000309366.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100156835.hg.1 | ENST00000536642.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100156835.hg.1 | ENST00000541388.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100156835.hg.1 | NM_004470.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100156835.hg.1 | TC1100007941.hg.1 | Clariom D TC |
