Assay Details
| Gene Symbol: PDCD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014976.1 | NP_055791.1 | 30 - 31 | 4647 | 57 | |
| XM_005269647.3 | XP_005269704.1 | 30 - 31 | 4687 | 57 | ||
| XM_011539538.2 | XP_011537840.1 | 30 - 31 | 4635 | 57 | ||
| XM_011539539.2 | XP_011537841.1 | 30 - 31 | 4635 | 57 | ||
| XM_011539540.1 | XP_011537842.1 | 29 - 30 | 4501 | 57 | ||
| GenBank mRNA | AK294971.1 | - | 3650 | 57 | ||
| D80007.1 | - | 30 - 31 | 4600 | 57 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000773.hg.1
Set Membership: |
|

PDCD11
Genomic Map