Assay Details
| Gene Symbol: RNASE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198235.2 | NP_937878.1 | 1 - 2 | 82 | 70 | |
| GenBank mRNA | CB993343.1 | - | 1 - 2 | 79 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000902.hg
Additional Information:
This assay is in the 5'UTR region of the NM_198235.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009773.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009773.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009773.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009773.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009773.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009773.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009773.hg.1 | TC14000902.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400161913.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400161913.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400161913.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400161913.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400161913.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400161913.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400161913.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400161913.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400161913.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400161913.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400161913.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400161913.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400161913.hg.1 | TC1400008606.hg.1 | Clariom D TC |

RNASE1
Genomic Map