Assay Details
| Gene Symbol: RNASEH2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006397.2 | NP_006388.2 | 1 - 2 | 189 | 72 | |
| GenBank mRNA | AK315327.1 | - | 1 - 2 | 219 | 72 | |
| AY363912.1 | - | 1 - 2 | 178 | 72 | ||
| BC011748.2 | - | 1 - 2 | 180 | 3160621 | 72 | |
| BG825340.1 | - | 1 - 2 | 193 | 72 | ||
| CR456841.1 | - | 1 - 2 | 129 | 72 | ||
| Z97029.1 | - | 1 - 2 | 189 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000229.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19003365.hg.1 | ENST00000221486.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003365.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003365.hg.1 | TC19000229.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000590121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000590121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000590121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000590121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000590121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000590279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000590279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000590279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000590279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000590279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000590279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003366.hg.1 | ENST00000221486.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003366.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003366.hg.1 | TC19000229.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000590121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000590121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000590121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000590121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000590121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000590279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000590279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000590279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000590279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000590279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000590279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007098.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900150981.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150981.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150981.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900150981.hg.1 | TC1900007098.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900150981.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150981.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150981.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900150984.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150984.hg.1 | ENST00000590121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150984.hg.1 | ENST00000590279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150984.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150984.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900150984.hg.1 | TC1900007098.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900150984.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150984.hg.1 | ENST00000590121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150984.hg.1 | ENST00000590279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150984.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150984.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900150986.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150986.hg.1 | ENST00000590121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150986.hg.1 | ENST00000590279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150986.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150986.hg.1 | NM_006397.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900150986.hg.1 | TC1900007098.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900150986.hg.1 | ENST00000221486.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150986.hg.1 | ENST00000590121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150986.hg.1 | ENST00000590279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150986.hg.1 | ENST00000593017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900150986.hg.1 | NM_006397.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 101370 | - | - |

RNASEH2A
Genomic Map