Assay Details
| Gene Symbol: HSD17B11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016245.4 | NP_057329.3 | 5 - 6 | 930 | 118 | |
| GenBank mRNA | AF273056.1 | - | 5 - 6 | 783 | 118 | |
| AK075348.1 | - | 5 - 6 | 814 | 118 | ||
| AY358553.1 | - | 5 - 6 | 797 | 118 | ||
| BC014327.1 | - | 5 - 6 | 796 | 3961721 | 118 | |
| BC016367.1 | - | 5 - 6 | 827 | 4133318 | 118 | |
| BC021673.1 | - | 5 - 6 | 801 | 3827935 | 118 | |
| BC036001.1 | - | 5 - 6 | 821 | 4715042 | 118 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04002945.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000502576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000502576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000502576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000502576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000502576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000502576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000508413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000508413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000508413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000508413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000512344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000512344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000512344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000512344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000512344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000512344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000513854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000513854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000513854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000513854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | ENST00000513854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019094.hg.1 | TC04002945.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019094.hg.1 | ENST00000358290.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019094.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019094.hg.1 | ENST00000507518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019094.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019098.hg.1 | TC04002945.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019098.hg.1 | ENST00000358290.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019098.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019098.hg.1 | ENST00000507518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019098.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190658.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190658.hg.1 | ENST00000502576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190658.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190658.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190658.hg.1 | ENST00000512344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190658.hg.1 | ENST00000513854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190658.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190658.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190658.hg.1 | ENST00000502576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190658.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190658.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190658.hg.1 | ENST00000512344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190658.hg.1 | ENST00000513854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190658.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190658.hg.1 | TC0400012945.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000502576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000502576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000502576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000502576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000502576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000502576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000508413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000508413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000508413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000508413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000512344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000512344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000512344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000512344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000512344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000512344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000513854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000513854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000513854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000513854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | ENST00000513854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012945.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190655.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190655.hg.1 | ENST00000502576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190655.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190655.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190655.hg.1 | ENST00000512344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190655.hg.1 | ENST00000513854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190655.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190655.hg.1 | ENST00000358290.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190655.hg.1 | ENST00000502576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190655.hg.1 | ENST00000507286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190655.hg.1 | ENST00000507518.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190655.hg.1 | ENST00000512344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190655.hg.1 | ENST00000513854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190655.hg.1 | NM_016245.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190655.hg.1 | TC0400012945.hg.1 | Clariom D TC |

HSD17B11
Genomic Map