Assay Details
| Gene Symbol: SNX29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032167.4 | NP_115543.3 | 21 | 7876 | 128 | |
| XM_011522741.2 | XP_011521043.1 | 22 | 8143 | 128 | ||
| GenBank mRNA | AK024473.1 | - | 1 | 4366 | 128 | |
| AL137333.1 | - | 1 | 1330 | 128 | ||
| BC013346.2 | - | 1 | 2207 | 128 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SNX29
Genomic Map