Assay Details
| Gene Symbol: RFX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002918.4 | NP_002909.4 | 10 - 11 | 1765 | 84 | |
| XM_011528165.2 | XP_011526467.1 | 11 - 12 | 2095 | 84 | ||
| XM_011528166.2 | XP_011526468.1 | 11 - 12 | 2031 | 84 | ||
| XM_011528167.2 | XP_011526469.1 | 11 - 12 | 2640 | 84 | ||
| XM_011528168.2 | XP_011526470.1 | 11 - 12 | 1801 | 84 | ||
| XM_011528169.2 | XP_011526471.1 | 10 - 11 | 1691 | 84 | ||
| XM_011528170.2 | XP_011526472.1 | 10 - 11 | 2269 | 84 | ||
| GenBank mRNA | A20498.1 | - | 10 - 11 | 1586 | 84 | |
| BC049826.1 | - | 10 - 11 | 1726 | 6043247 | 84 | |
| X58964.1 | - | 10 - 11 | 1586 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001227.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19017253.hg.1 | ENST00000254325.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017253.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017253.hg.1 | TC19001227.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000586913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000586913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180137.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180137.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180137.hg.1 | TC1900009795.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900180137.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180137.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000586913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000586913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |

RFX1
Genomic Map