Assay Details
| Gene Symbol: RFX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002918.4 | NP_002909.4 | 2 - 3 | 589 | 72 | |
| XM_011528165.2 | XP_011526467.1 | 2 - 3 | 589 | 72 | ||
| XM_011528166.2 | XP_011526468.1 | 2 - 3 | 525 | 72 | ||
| XM_011528167.2 | XP_011526469.1 | 2 - 3 | 1134 | 72 | ||
| XM_011528168.2 | XP_011526470.1 | 2 - 3 | 589 | 72 | ||
| GenBank mRNA | A20498.1 | - | 2 - 3 | 410 | 72 | |
| AK309824.1 | - | 2 - 3 | 588 | 72 | ||
| BC049826.1 | - | 2 - 3 | 550 | 6043247 | 72 | |
| X58964.1 | - | 2 - 3 | 410 | 72 | ||
More Information
Set Membership: |
|

RFX1
Genomic Map