Assay Details
| Gene Symbol: RFX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002918.4 | NP_002909.4 | 6 - 7 | 1010 | 109 | |
| XM_011528165.2 | XP_011526467.1 | 6 - 7 | 1010 | 109 | ||
| XM_011528166.2 | XP_011526468.1 | 6 - 7 | 946 | 109 | ||
| XM_011528167.2 | XP_011526469.1 | 6 - 7 | 1555 | 109 | ||
| XM_011528168.2 | XP_011526470.1 | 6 - 7 | 1010 | 109 | ||
| XM_011528169.2 | XP_011526471.1 | 5 - 6 | 606 | 109 | ||
| XM_011528170.2 | XP_011526472.1 | 5 - 6 | 1184 | 109 | ||
| GenBank mRNA | A20498.1 | - | 6 - 7 | 831 | 109 | |
| AK309824.1 | - | 6 - 7 | 1009 | 109 | ||
| BC049826.1 | - | 6 - 7 | 971 | 6043247 | 109 | |
| X58964.1 | - | 6 - 7 | 831 | 109 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001227.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19017259.hg.1 | ENST00000254325.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017259.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017259.hg.1 | TC19001227.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017258.hg.1 | ENST00000254325.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017258.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017258.hg.1 | TC19001227.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000586913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000586913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180143.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180143.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180143.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180143.hg.1 | TC1900009795.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900180143.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180143.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180143.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000586913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000586913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000588885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | ENST00000589937.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009795.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180142.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180142.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180142.hg.1 | NM_002918.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180142.hg.1 | TC1900009795.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900180142.hg.1 | ENST00000254325.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180142.hg.1 | ENST00000589239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180142.hg.1 | NM_002918.4 | Refseq |

RFX1
Genomic Map