Assay Details
| Gene Symbol: MUC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164462.1 | NP_001157934.1 | 8 - 9 | 15639 | 69 | |
| GenBank mRNA | AF147790.1 | - | 7 - 8 | 1391 | 69 | |
| AK024856.1 | - | 7 - 8 | 1146 | 69 | ||
| BC127877.1 | - | 7 - 8 | 1409 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000639.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000305119.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000305119.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000305119.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000305119.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000305119.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000467414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000467414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000467414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000467414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000467414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000467414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000473098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000473098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000473098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000473098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000473098.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000474482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000474482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000474482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000474482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000474482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000474482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009964.hg.1 | TC07000639.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009964.hg.1 | ENST00000379442.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009964.hg.1 | ENST00000536621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07009964.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700160634.hg.1 | ENST00000305119.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160634.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160634.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160634.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700160634.hg.1 | ENST00000305119.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160634.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160634.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700160634.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700160634.hg.1 | TC0700008576.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000305119.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000305119.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000305119.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000305119.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000305119.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000379442.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000467414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000467414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000467414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000467414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000467414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000467414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000473098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000473098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000473098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000473098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000473098.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000474482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000474482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000474482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000474482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000474482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000474482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | ENST00000536621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008576.hg.1 | NM_001164462.1 | Refseq |

MUC12
Genomic Map