Assay Details
| Gene Symbol: ICAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000201.2 | NP_000192.2 | 5 - 6 | 1500 | 90 | |
| GenBank mRNA | AF340038.1 | - | 5 - 6 | 1181 | 90 | |
| AF340039.1 | - | 5 - 6 | 1169 | 90 | ||
| AK130659.1 | - | 5 - 6 | 1221 | 90 | ||
| AK298054.1 | - | 5 - 6 | 1107 | 90 | ||
| AK298983.1 | - | 5 - 6 | 933 | 90 | ||
| AK301412.1 | - | 3 - 4 | 582 | 90 | ||
| AK312636.1 | - | 5 - 6 | 1221 | 90 | ||
| BC015969.2 | - | 5 - 6 | 1252 | 3506766 | 90 | |
| BT006854.1 | - | 5 - 6 | 1181 | 90 | ||
| J03132.1 | - | 5 - 6 | 1238 | 90 | ||
| M24283.1 | - | 5 - 6 | 1252 | 90 | ||
| M55038.1 | - | 2 - 3 | 413 | 90 | ||
| X06990.1 | - | 5 - 6 | 1193 | 90 | ||
More Information
Set Membership: |
|

ICAM1
Genomic Map