Assay Details
| Gene Symbol: SPAG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004890.2 | NP_004881.2 | 2 - 3 | 186 | 99 | |
| XM_006721600.2 | XP_006721663.1 | 2 - 3 | 811 | 99 | ||
| GenBank mRNA | AF047437.1 | - | 2 - 3 | 163 | 99 | |
| AK123065.1 | - | 965 | 99 | |||
| AK312174.1 | - | 2 - 3 | 162 | 99 | ||
| BC011934.2 | - | 2 - 3 | 155 | 4330461 | 99 | |
| BP308951.1 | - | 2 - 3 | 185 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006721600.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000570341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000570341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000571023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000571023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000571023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000570341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000570341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000571023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000571023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000571023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173463.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173463.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173463.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173463.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173463.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173463.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173463.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173463.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173463.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173463.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173463.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173463.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173463.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173463.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173463.hg.1 | TC1700009540.hg.1 | Clariom D TC |

SPAG7
Genomic Map