Assay Details
| Gene Symbol: SPAG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004890.2 | NP_004881.2 | 3 - 4 | 277 | 89 | |
| XM_006721600.2 | XP_006721663.1 | 3 - 4 | 902 | 89 | ||
| GenBank mRNA | AF047437.1 | - | 3 - 4 | 254 | 89 | |
| AK123065.1 | - | 1056 | 89 | |||
| AK312174.1 | - | 3 - 4 | 253 | 89 | ||
| BC011934.2 | - | 3 - 4 | 246 | 4330461 | 89 | |
| BP308951.1 | - | 3 - 4 | 276 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000570341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000570341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000571023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000571023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000571023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000570341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000570341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000571023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000571023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000571023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | ENST00000575784.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009540.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173461.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173461.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173461.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173461.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173461.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173461.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173461.hg.1 | ENST00000206020.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173461.hg.1 | ENST00000572148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173461.hg.1 | ENST00000573366.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173461.hg.1 | ENST00000573805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173461.hg.1 | ENST00000575142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700173461.hg.1 | NM_004890.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700173461.hg.1 | TC1700009540.hg.1 | Clariom D TC |

SPAG7
Genomic Map