Assay Details
| Gene Symbol: CAMTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171167.1 | NP_001164638.1 | 20 - 21 | 3757 | 76 | |
| NM_001171168.1 | NP_001164639.1 | 19 - 20 | 3685 | 76 | ||
| NM_015099.3 | NP_055914.2 | 20 - 21 | 3413 | 76 | ||
| XM_006721478.3 | XP_006721541.1 | 20 - 21 | 4060 | 76 | ||
| XM_006721481.3 | XP_006721544.1 | 19 - 20 | 3973 | 76 | ||
| XM_006721482.3 | XP_006721545.1 | 18 - 19 | 3106 | 76 | ||
| XM_011523746.2 | XP_011522048.1 | 20 - 21 | 3420 | 76 | ||
| XM_011523747.2 | XP_011522049.1 | 20 - 21 | 3404 | 76 | ||
| GenBank mRNA | AB020716.1 | - | 20 - 21 | 3374 | 76 | |
| AL831849.1 | - | 19 - 20 | 3685 | 76 | ||
| BC016163.1 | - | 7 - 8 | 1032 | 76 | ||
| BC032381.1 | - | 4 - 5 | 428 | 76 | ||
| BC136534.1 | - | 19 - 20 | 3342 | 9020524 | 76 | |
| BC142695.1 | - | 20 - 21 | 3404 | 40148717 | 76 | |
| BC144232.1 | - | 18 - 19 | 3270 | 9052752 | 76 | |
| BC167148.1 | - | 20 - 21 | 3374 | 8862515 | 76 | |
| BC167149.1 | - | 20 - 21 | 3374 | 8862516 | 76 | |
| BC167151.1 | - | 20 - 21 | 3374 | 8862527 | 76 | |
| BC167160.1 | - | 20 - 21 | 3374 | 8862594 | 76 | |
More Information
Set Membership: |
|

CAMTA2
Genomic Map