Assay Details
| Gene Symbol: ELAVL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011517783.2 | XP_011516085.1 | 2 - 3 | 525 | 105 | |
| XM_017014417.1 | XP_016869906.1 | 2 - 3 | 525 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011517783.2, XM_017014417.1 transcripts.
Set Membership: |
|

ELAVL2
Genomic Map