Assay Details
| Gene Symbol: ANKRD13D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011544988.2 | XP_011543290.1 | 3 - 4 | 323 | 83 | |
| GenBank mRNA | AK027313.1 | - | 1 - 2 | 991 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK027313.1, XM_011544988.2 transcripts.
Set Membership: |
|

ANKRD13D
Genomic Map