Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021447.hg.1 |
ENST00000244906.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021447.hg.1 |
ENST00000536405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021447.hg.1 |
NR_026882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021447.hg.1 |
TC11001856.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013169.hg.1 |
ENST00000244906.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013169.hg.1 |
ENST00000244906.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013169.hg.1 |
ENST00000244906.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013169.hg.1 |
ENST00000244906.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013169.hg.1 |
ENST00000244906.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013169.hg.1 |
ENST00000244906.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013169.hg.1 |
NR_026882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013169.hg.1 |
NR_026882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013169.hg.1 |
NR_026882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013169.hg.1 |
NR_026882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013169.hg.1 |
NR_026882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013169.hg.1 |
NR_026882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204194.hg.1 |
ENST00000244906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204194.hg.1 |
NR_026882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204194.hg.1 |
ENST00000244906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204194.hg.1 |
NR_026882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204194.hg.1 |
TC1100013169.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100013169.hg.1 |
ENST00000244906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013169.hg.1 |
ENST00000244906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013169.hg.1 |
ENST00000244906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013169.hg.1 |
ENST00000244906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013169.hg.1 |
ENST00000244906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013169.hg.1 |
ENST00000244906.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013169.hg.1 |
NR_026882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013169.hg.1 |
NR_026882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013169.hg.1 |
NR_026882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013169.hg.1 |
NR_026882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013169.hg.1 |
NR_026882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013169.hg.1 |
NR_026882.1 |
Refseq |