Assay Details
| Gene Symbol: ABCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000392.4 | NP_000383.1 | 14 - 15 | 2151 | 66 | |
| XM_006717630.3 | XP_006717693.1 | 9 - 10 | 1459 | 66 | ||
| XM_006717631.3 | XP_006717694.1 | 14 - 15 | 2128 | 66 | ||
| XM_011539291.2 | XP_011537593.1 | 14 - 15 | 2127 | 66 | ||
| XM_017015675.1 | XP_016871164.1 | 14 - 15 | 2128 | 66 | ||
| GenBank mRNA | BC136419.1 | - | 14 - 15 | 1939 | 9020407 | 66 |
| U49248.1 | - | 14 - 15 | 2005 | 66 | ||
| U63970.1 | - | 14 - 15 | 1941 | 66 | ||
| X96395.2 | - | 14 - 15 | 1941 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transport
drug transmembrane transport
response to oxidative stress
response to heat
bilirubin transport
prostaglandin transport
cellular chloride ion homeostasis
response to methotrexate
response to estrogen
response to arsenic-containing substance
response to steroid hormone
transmembrane transport
thyroid hormone transport
anion transmembrane transport
protein binding
ATP binding
organic anion transmembrane transporter activity
bilirubin transmembrane transporter activity
protein domain specific binding
ATPase activity, coupled to transmembrane movement of substances
anion transmembrane-transporting ATPase activity
transport
drug transmembrane transport
response to oxidative stress
response to heat
bilirubin transport
prostaglandin transport
cellular chloride ion homeostasis
response to methotrexate
response to estrogen
response to arsenic-containing substance
response to steroid hormone
transmembrane transport
thyroid hormone transport
anion transmembrane transport
protein binding
ATP binding
organic anion transmembrane transporter activity
bilirubin transmembrane transporter activity
protein domain specific binding
ATPase activity, coupled to transmembrane movement of substances
anion transmembrane-transporting ATPase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000496621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000496621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000496621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000496621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | ENST00000496621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158852.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158852.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158852.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158852.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158852.hg.1 | TC1000008628.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000370449.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000496621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000496621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000496621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000496621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | ENST00000496621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008628.hg.1 | NM_000392.4 | Refseq |

ABCC2
Genomic Map