Assay Details
| Gene Symbol: AS3MT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020682.3 | NP_065733.2 | 3 - 4 | 312 | 79 | |
| GenBank mRNA | AF226730.1 | - | 3 - 4 | 249 | 79 | |
| AK057833.1 | - | 3 - 4 | 331 | 79 | ||
| AK225972.1 | - | 3 - 4 | 331 | 79 | ||
| AK309737.1 | - | 3 - 4 | 244 | 79 | ||
| BC119637.1 | - | 3 - 4 | 216 | 79 | ||
| BC119638.2 | - | 3 - 4 | 216 | 79 | ||
| DA368970.1 | - | 3 - 4 | 312 | 79 | ||
| Gene Symbol: BORCS7-ASMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037644.1 | 7 - 8 | 577 | 79 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10002948.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009129.hg.1 | TC10002948.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009129.hg.1 | ENST00000369880.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009129.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186250.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186250.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186250.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186250.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186250.hg.1 | TC1000012497.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186223.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186223.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186223.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186223.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186223.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186223.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186223.hg.1 | TC1000012496.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000369880.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | ENST00000615257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012496.hg.1 | NM_020682.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |

AS3MT
Genomic Map