Assay Details
| Gene Symbol: BORCS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136200.1 | NP_001129672.1 | 2 - 3 | 276 | 137 | |
| NM_144591.3 | NP_653192.2 | 2 - 3 | 276 | 137 | ||
| GenBank mRNA | AK098071.1 | - | 2 - 3 | 224 | 137 | |
| AK311742.1 | - | 2 - 3 | 206 | 137 | ||
| AL832477.1 | - | 2 - 3 | 264 | 137 | ||
| BC015994.1 | - | 2 - 3 | 228 | 4698381 | 137 | |
| DA006534.1 | - | 2 - 3 | 277 | 137 | ||
| Gene Symbol: BORCS7-ASMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037644.1 | 2 - 3 | 276 | 137 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10002947.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10009118.hg.1 | TC10002947.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009118.hg.1 | ENST00000339834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009118.hg.1 | ENST00000369883.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009118.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009118.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000369883.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000369883.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000369883.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000369883.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000369883.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000369883.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000478833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000478833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | ENST00000478833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012495.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012497.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186245.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186245.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186245.hg.1 | ENST00000299353.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186245.hg.1 | NR_037644.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186245.hg.1 | TC1000012497.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000369883.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000369883.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000369883.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000369883.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000369883.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000369883.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000478833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000478833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | ENST00000478833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012495.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186209.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186209.hg.1 | ENST00000369883.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186209.hg.1 | ENST00000478833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186209.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186209.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186209.hg.1 | ENST00000339834.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186209.hg.1 | ENST00000369883.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186209.hg.1 | ENST00000478833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186209.hg.1 | NM_001136200.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186209.hg.1 | NM_144591.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186209.hg.1 | TC1000012495.hg.1 | Clariom D TC |

BORCS7
Genomic Map