Assay Details
| Gene Symbol: ARHGEF38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242729.1 | NP_001229658.1 | 8 - 9 | 1265 | 92 | |
| XM_006714244.3 | XP_006714307.1 | 6 - 7 | 625 | 92 | ||
| XM_011532050.2 | XP_011530352.1 | 7 - 8 | 814 | 92 | ||
| XM_011532051.2 | XP_011530353.1 | 8 - 9 | 1280 | 92 | ||
| GenBank mRNA | DT932593.1 | - | 5 - 6 | 576 | 92 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000544.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007912.hg.1 | TC04000544.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007912.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007912.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007912.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188724.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188724.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188724.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188724.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188724.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188724.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188724.hg.1 | TC0400012829.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |

ARHGEF38
Genomic Map