Assay Details
| Gene Symbol: ARHGEF38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242729.1 | NP_001229658.1 | 11 - 12 | 1840 | 93 | |
| XM_006714244.3 | XP_006714307.1 | 9 - 10 | 1200 | 93 | ||
| XM_011532050.2 | XP_011530352.1 | 10 - 11 | 1389 | 93 | ||
| GenBank mRNA | AK125951.1 | - | 2 - 3 | 318 | 93 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000544.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04007915.hg.1 | TC04000544.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007915.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007915.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007915.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000265154.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000506828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012829.hg.1 | NM_017700.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188727.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188727.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188727.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188727.hg.1 | ENST00000420470.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188727.hg.1 | ENST00000508036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188727.hg.1 | NM_001242729.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188727.hg.1 | TC0400012829.hg.1 | Clariom D TC |

ARHGEF38
Genomic Map