Assay Details
| Gene Symbol: PATL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152716.2 | NP_689929.2 | 2 - 3 | 269 | 97 | |
| XM_005273833.4 | XP_005273890.1 | 2 - 3 | 1391 | 97 | ||
| XM_005273834.1 | XP_005273891.1 | 2 - 3 | 271 | 97 | ||
| XM_017017339.1 | XP_016872828.1 | 2 - 3 | 271 | 97 | ||
| GenBank mRNA | AK127943.1 | - | 2 - 3 | 229 | 97 | |
| AL831992.2 | - | 2 - 3 | 210 | 97 | ||
| DC409866.1 | - | 2 - 3 | 269 | 97 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001829.hg
Additional Information:
This assay is in the 5'UTR region of the XM_017017339.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
deadenylation-dependent decapping of nuclear-transcribed mRNA
cytoplasmic mRNA processing body assembly
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
RNA binding
protein binding
poly(U) RNA binding
poly(G) binding
poly(A) RNA binding
deadenylation-dependent decapping of nuclear-transcribed mRNA
cytoplasmic mRNA processing body assembly
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
RNA binding
protein binding
poly(U) RNA binding
poly(G) binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900010685.hg.1 | ENST00000597336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010685.hg.1 | ENST00000597336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021075.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021075.hg.1 | ENST00000428532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021075.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021075.hg.1 | TC11001829.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021076.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021076.hg.1 | ENST00000428532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021076.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021076.hg.1 | TC11001829.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008867.hg.1 | ENST00000449048.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008867.hg.1 | ENST00000449048.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000531919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000531919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | ENST00000531919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021074.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021074.hg.1 | ENST00000428532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021074.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021074.hg.1 | TC11001829.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021078.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021078.hg.1 | TC11001829.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100182825.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182825.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182825.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182825.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182825.hg.1 | TC1100010981.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100182826.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182826.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182826.hg.1 | TC1100010981.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100182823.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182823.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182823.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182823.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182823.hg.1 | TC1100010981.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100182822.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182822.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182822.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182822.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182822.hg.1 | TC1100010981.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1900010685.hg.1 | ENST00000597336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010685.hg.1 | ENST00000597336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008867.hg.1 | ENST00000449048.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008867.hg.1 | ENST00000449048.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000300146.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000531919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000531919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | ENST00000531919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010981.hg.1 | NM_152716.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 331519 | - | - |

PATL1
Genomic Map