Assay Details
| Gene Symbol: PATL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005273834.1 | XP_005273891.1 | 6 - 7 | 864 | 82 | |
| GenBank mRNA | AK127943.1 | - | 6 - 7 | 822 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PATL1
Genomic Map