Assay Details
| Gene Symbol: TNKS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025235.3 | NP_079511.1 | 20 - 21 | 2991 | 78 | |
| XM_005270185.4 | XP_005270242.2 | 20 - 21 | 3031 | 78 | ||
| XM_011540213.1 | XP_011538515.1 | 20 - 21 | 3022 | 78 | ||
| XM_011540215.2 | XP_011538517.1 | 12 - 13 | 3216 | 78 | ||
| XM_017016696.1 | XP_016872185.1 | 20 - 21 | 2968 | 78 | ||
| XM_017016697.1 | XP_016872186.1 | 20 - 21 | 2984 | 78 | ||
| XM_017016698.1 | XP_016872187.1 | 20 - 21 | 2695 | 78 | ||
| XM_017016699.1 | XP_016872188.1 | 20 - 21 | 2975 | 78 | ||
| XM_017016700.1 | XP_016872189.1 | 20 - 21 | 2581 | 78 | ||
| GenBank mRNA | AF264912.1 | - | 20 - 21 | 2694 | 78 | |
| AF305081.1 | - | 20 - 21 | 2993 | 78 | ||
| AF309033.1 | - | 20 - 21 | 2945 | 78 | ||
| AF329696.1 | - | 20 - 21 | 2833 | 78 | ||
| AF342982.1 | - | 20 - 21 | 2694 | 78 | ||
| AF438201.1 | - | 20 - 21 | 3073 | 78 | ||
| AK023746.1 | - | 2 - 3 | 185 | 78 | ||
| AK314612.1 | - | 20 - 21 | 2959 | 78 | ||
| AL833676.1 | - | 6 - 7 | 1029 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000654.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK023746.1 transcripts.
Set Membership: |
|

TNKS2
Genomic Map