Assay Details
| Gene Symbol: IL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000572.2 | NP_000563.1 | 1 - 2 | 224 | 98 | |
| GenBank mRNA | AF043333.1 | - | 1 - 2 | 111 | 98 | |
| AY029171.1 | - | 1 - 2 | 165 | 98 | ||
| BC104252.1 | - | 1 - 2 | 221 | 40035920 | 98 | |
| BC104253.1 | - | 1 - 2 | 221 | 40035922 | 98 | |
| CR541993.1 | - | 1 - 2 | 165 | 98 | ||
| CR542028.1 | - | 1 - 2 | 165 | 98 | ||
| M57627.1 | - | 1 - 2 | 195 | 98 | ||
More Information
Set Membership: |
|

IL10
Genomic Map