Assay Details
| Gene Symbol: SHD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020209.3 | NP_064594.3 | 2 - 3 | 1864 | 87 | |
| GenBank mRNA | AK055673.1 | - | 2 - 3 | 803 | 87 | |
| AK056268.1 | - | 2 - 3 | 1865 | 87 | ||
| AL390078.1 | - | 2 - 3 | 143 | 87 | ||
| BC007206.1 | - | 2 - 3 | 626 | 3628187 | 87 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000085.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19001364.hg.1 | ENST00000221852.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001364.hg.1 | ENST00000543264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001364.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19001364.hg.1 | TC19000085.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000593383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000593383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000593383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000593383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000593383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000593383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000597466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000597466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000597466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000597466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000600475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000600475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | ENST00000600475.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146863.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146863.hg.1 | ENST00000593383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146863.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146863.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146863.hg.1 | TC1900006684.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900146863.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146863.hg.1 | ENST00000593383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146863.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146863.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146862.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146862.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146862.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900146862.hg.1 | TC1900006684.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900146862.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146862.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900146862.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000543264.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000593383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000593383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000593383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000593383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000593383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000593383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000597466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000597466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000597466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000597466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000599689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000600475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000600475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | ENST00000600475.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | NM_020209.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006684.hg.1 | NM_020209.3 | Refseq |

SHD
Genomic Map