Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011232.hg.1 |
ENST00000331442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011232.hg.1 |
ENST00000331442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011232.hg.1 |
ENST00000331442.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011232.hg.1 |
NM_005322.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011232.hg.1 |
NM_005322.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011232.hg.1 |
NM_005322.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011232.hg.1 |
NM_005322.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011232.hg.1 |
NM_005322.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016358.hg.1 |
TC06001426.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016358.hg.1 |
ENST00000331442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016358.hg.1 |
NM_005322.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016355.hg.1 |
TC06001426.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016355.hg.1 |
ENST00000331442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016355.hg.1 |
NM_005322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175730.hg.1 |
ENST00000331442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175730.hg.1 |
NM_005322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175730.hg.1 |
ENST00000331442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175730.hg.1 |
NM_005322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175730.hg.1 |
TC0600011232.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600175733.hg.1 |
NM_005322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175733.hg.1 |
NM_005322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175733.hg.1 |
TC0600011232.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011232.hg.1 |
ENST00000331442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011232.hg.1 |
ENST00000331442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011232.hg.1 |
ENST00000331442.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011232.hg.1 |
NM_005322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011232.hg.1 |
NM_005322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011232.hg.1 |
NM_005322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011232.hg.1 |
NM_005322.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011232.hg.1 |
NM_005322.2 |
Refseq |