Assay Details
| Gene Symbol: PAXBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013329.3 | NP_037461.2 | 8 - 9 | 1693 | 97 | |
| NM_016631.3 | NP_057715.2 | 8 - 9 | 1693 | 97 | ||
| XM_006724066.1 | XP_006724129.1 | 8 - 9 | 1622 | 97 | ||
| XM_011529804.2 | XP_011528106.1 | 8 - 9 | 1622 | 97 | ||
| GenBank mRNA | AF231920.1 | - | 7 - 8 | 1110 | 97 | |
| AJ279080.1 | - | 8 - 9 | 1519 | 97 | ||
| AY033903.1 | - | 8 - 9 | 1503 | 97 | ||
| AY033904.1 | - | 8 - 9 | 1503 | 97 | ||
| BC005213.1 | - | 1 - 2 | 119 | 97 | ||
More Information
Set Membership: |
|

PAXBP1
Genomic Map