Assay Details
Gene Symbol: PFN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199346.2 | NP_955378.1 | 2 - 3 | 490 | 61 | |
XM_005264292.2 | XP_005264349.1 | 3 - 4 | 559 | 61 | ||
XM_006712007.3 | XP_006712070.1 | 3 - 4 | 301 | 61 | ||
XM_011532823.2 | XP_011531125.1 | 3 - 4 | 518 | 61 | ||
GenBank mRNA | AK292601.1 | - | 2 - 3 | 490 | 61 | |
BC029523.1 | - | 2 - 3 | 294 | 5297707 | 61 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001646.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02026245.hg.1 | TC02001646.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02026245.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026245.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026245.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02026247.hg.1 | TC02001646.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02026247.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026247.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026247.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02026246.hg.1 | TC02001646.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02026246.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026246.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026246.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02026251.hg.1 | TC02001646.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02026251.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026251.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026251.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02026249.hg.1 | TC02001646.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02026249.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026249.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02026248.hg.1 | TC02001646.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02026248.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02026248.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187018.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187018.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187018.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187018.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187018.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187018.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187018.hg.1 | TC0200011976.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200187017.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187017.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187017.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187017.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187017.hg.1 | TC0200011976.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200187016.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187016.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187016.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187016.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187016.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187016.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187016.hg.1 | TC0200011976.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200187022.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187022.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187022.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187022.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187022.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187022.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187022.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187022.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187022.hg.1 | TC0200011976.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011976.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187021.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187021.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187021.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187021.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187021.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187021.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187021.hg.1 | ENST00000465360.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187021.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187021.hg.1 | TC0200011976.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200187023.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187023.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187023.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187023.hg.1 | ENST00000313213.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187023.hg.1 | ENST00000436622.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200187023.hg.1 | NM_199346.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200187023.hg.1 | TC0200011976.hg.1 | Clariom D TC |
