Assay Details
| Gene Symbol: B4GALNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159387.1 | NP_001152859.1 | 6 - 7 | 707 | 67 | |
| NM_001159388.1 | NP_001152860.1 | 6 - 7 | 720 | 67 | ||
| NM_153446.2 | NP_703147.2 | 6 - 7 | 922 | 67 | ||
| XM_011524314.2 | XP_011522616.1 | 6 - 7 | 1075 | 67 | ||
| XM_017024173.1 | XP_016879662.1 | 6 - 7 | 769 | 67 | ||
| GenBank mRNA | AF510036.1 | - | 6 - 7 | 922 | 67 | |
| AJ517770.1 | - | 6 - 7 | 922 | 67 | ||
| AJ517771.1 | - | 6 - 7 | 704 | 67 | ||
| AK302876.1 | - | 6 - 7 | 720 | 67 | ||
| BC113675.1 | - | 6 - 7 | 916 | 8322727 | 67 | |
| BC113677.1 | - | 6 - 7 | 916 | 8322729 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000638.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000575159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007977.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007977.hg.1 | ENST00000393354.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007977.hg.1 | ENST00000504681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007977.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007977.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007977.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007977.hg.1 | TC17000638.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700160594.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160594.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160594.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160594.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160594.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160594.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160594.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160594.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160594.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160594.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160594.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160594.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160594.hg.1 | TC1700008191.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000575159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |

B4GALNT2
Genomic Map