Assay Details
| Gene Symbol: B4GALNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159387.1 | NP_001152859.1 | 8 - 9 | 985 | 83 | |
| NM_001159388.1 | NP_001152860.1 | 8 - 9 | 998 | 83 | ||
| NM_153446.2 | NP_703147.2 | 8 - 9 | 1200 | 83 | ||
| XM_017024173.1 | XP_016879662.1 | 8 - 9 | 1047 | 83 | ||
| GenBank mRNA | AF510036.1 | - | 8 - 9 | 1200 | 83 | |
| AJ517770.1 | - | 8 - 9 | 1200 | 83 | ||
| AJ517771.1 | - | 8 - 9 | 982 | 83 | ||
| AK302876.1 | - | 8 - 9 | 998 | 83 | ||
| BC113675.1 | - | 8 - 9 | 1194 | 8322727 | 83 | |
| BC113677.1 | - | 8 - 9 | 1194 | 8322729 | 83 | |
| S83275.1 | - | 2 - 3 | 262 | 83 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000638.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17007979.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007979.hg.1 | ENST00000393354.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007979.hg.1 | ENST00000504681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007979.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007979.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007979.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17007979.hg.1 | TC17000638.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | ENST00000575159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160598.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160598.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160598.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160598.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160598.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160598.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160598.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160598.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160598.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700160598.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160598.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160598.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700160598.hg.1 | TC1700008191.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000300404.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000393354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000504681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | ENST00000575159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159387.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_001159388.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008191.hg.1 | NM_153446.2 | Refseq |

B4GALNT2
Genomic Map