Assay Details
| Gene Symbol: ALAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000688.5 | NP_000679.1 | 2 - 3 | 344 | 74 | |
| GenBank mRNA | AY260745.1 | - | 2 - 3 | 283 | 74 | |
| BC011798.2 | - | 2 - 3 | 242 | 4138141 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY260745.1, BC011798.2, NM_000688.5 transcripts.
Set Membership: |
|

ALAS1
Genomic Map