Assay Details
| Gene Symbol: BLZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320973.1 | NP_001307902.1 | 4 - 5 | 1180 | 77 | |
| NM_003666.3 | NP_003657.1 | 4 - 5 | 1180 | 77 | ||
| XM_005245561.4 | XP_005245618.1 | 4 - 5 | 1087 | 77 | ||
| XM_011510092.2 | XP_011508394.1 | 4 - 5 | 1083 | 77 | ||
| GenBank mRNA | AK223533.1 | - | 4 - 5 | 1042 | 77 | |
| AK291725.1 | - | 4 - 5 | 847 | 77 | ||
| AK307151.1 | - | 5 - 6 | 688 | 77 | ||
| BC020716.1 | - | 4 - 5 | 829 | 4723421 | 77 | |
| U79751.1 | - | 4 - 5 | 1069 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001475.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
regulation of transcription from RNA polymerase II promoter
Golgi organization
cell proliferation
vesicle-mediated transport
Golgi to plasma membrane protein transport
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
enzyme binding
ubiquitin protein ligase binding
regulation of cell growth
regulation of transcription from RNA polymerase II promoter
Golgi organization
cell proliferation
vesicle-mediated transport
Golgi to plasma membrane protein transport
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
enzyme binding
ubiquitin protein ligase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400009528.hg.1 | ENST00000553776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009528.hg.1 | ENST00000553776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009528.hg.1 | ENST00000553776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009528.hg.1 | ENST00000553776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022867.hg.1 | TC01001475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022867.hg.1 | ENST00000329281.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022867.hg.1 | ENST00000367808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022867.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022868.hg.1 | TC01001475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022868.hg.1 | ENST00000329281.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022868.hg.1 | ENST00000367808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022868.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022866.hg.1 | TC01001475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022866.hg.1 | ENST00000329281.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022866.hg.1 | ENST00000367808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022866.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177623.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177623.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177623.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177623.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177623.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177623.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177623.hg.1 | TC0100010546.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100177621.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177621.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177621.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177621.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177621.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177621.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177621.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177621.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177621.hg.1 | TC0100010546.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100177622.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177622.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177622.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177622.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177622.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177622.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177622.hg.1 | TC0100010546.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009528.hg.1 | ENST00000553776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009528.hg.1 | ENST00000553776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009528.hg.1 | ENST00000553776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009528.hg.1 | ENST00000553776.1 | Ensembl |

BLZF1
Genomic Map